Anti-PAFAH1B3 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3171AEA 350 EUR
ABGEAT3171A
Anti-PAFAH1B3 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PAFAH1B3 (platelet-activating factor acetylhydrolase 1b, catalytic subunit 3 (29kDa))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR