Anti-PAK1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3173AEA 350 EUR
ABGEAT3173A
Anti-PAK1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PAK1 (p21 protein (Cdc42/Rac)-activated kinase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR