Anti-PCMT1 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT3234AEA 350 EUR
ABGEAT3234A
Anti-PCMT1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PCMT1 (protein-L-isoaspartate (D-aspartate) O-methyltransferase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human