Anti-PCMT1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3235AEA 350 EUR
ABGEAT3235A
Anti-PCMT1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PCMT1 (protein-L-isoaspartate (D-aspartate) O-methyltransferase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR