Anti-PIN1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3312AEA 350 EUR
ABGEAT3312A
Anti-PIN1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PIN1 (peptidylprolyl cis/trans isomerase, NIMA-interacting 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR