Anti-PLA2G1B Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3328AEA 350 EUR
ABGEAT3328A
Anti-PLA2G1B Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PLA2G1B (phospholipase A2, group IB (pancreas))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR