Anti-PLS1 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT3347AEA 350 EUR
ABGEAT3347A
Anti-PLS1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PLS1 (plastin 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human