Anti-POLR2I Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3377AEA 350 EUR
ABGEAT3377A
Anti-POLR2I Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: POLR2I (polymerase (RNA) II (DNA directed) polypeptide I, 14.5kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR