Anti-PRKAB1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3427AEA 350 EUR
ABGEAT3427A
Anti-PRKAB1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PRKAB1 (protein kinase, AMP-activated, beta 1 non-catalytic subunit)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR