Anti-PSMB4 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT3460AEA 350 EUR
ABGEAT3460A
Anti-PSMB4 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PSMB4 (proteasome (prosome, macropain) subunit, beta type, 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human