Anti-PSMD10 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3468AEA 350 EUR
ABGEAT3468A
Anti-PSMD10 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PSMD10 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 10)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR