Anti-PTGIR Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3484AEA 350 EUR
ABGEAT3484A
Anti-PTGIR Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PTGIR (prostaglandin I2 (prostacyclin) receptor (IP))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR