Anti-PTRF Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT3498AEA 350 EUR
ABGEAT3498A
Anti-PTRF Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PTRF (polymerase I and transcript release factor)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human