Anti-RAB7B Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT3533AEA 350 EUR
ABGEAT3533A
Anti-RAB7B Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RAB7B (RAB7B, member RAS oncogene family)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human