Anti-RALGPS2 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3557AEA 350 EUR
ABGEAT3557A
Anti-RALGPS2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RALGPS2 (Ral GEF with PH domain and SH3 binding motif 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR