Anti-RASEF Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT3577AEA 350 EUR
ABGEAT3577A
Anti-RASEF Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RASEF (RAS and EF-hand domain containing)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human