Anti-RASGEF1C Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT3578AEA 350 EUR
ABGEAT3578A
Anti-RASGEF1C Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RASGEF1C (RasGEF domain family, member 1C)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human