Anti-REG1A Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT3614AEA 350 EUR
ABGEAT3614A
Anti-REG1A Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: REG1A (regenerating islet-derived 1 alpha)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human