Anti-RFFL Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3622AEA 350 EUR
ABGEAT3622A
Anti-RFFL Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RFFL (ring finger and FYVE-like domain containing E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR