Anti-RFNG Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3623AEA 350 EUR
ABGEAT3623A
Anti-RFNG Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RFNG (RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR