Anti-RNASEH2A Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3653AEA 350 EUR
ABGEAT3653A
Anti-RNASEH2A Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RNASEH2A (ribonuclease H2, subunit A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR