Anti-RNF139 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT3665AEA 350 EUR
ABGEAT3665A
Anti-RNF139 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RNF139 (ring finger protein 139)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human