Anti-RPL10 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT3700AEA 350 EUR
ABGEAT3700A
Anti-RPL10 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RPL10 (ribosomal protein L10)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human