Anti-RPL14 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT3703AEA 350 EUR
ABGEAT3703A
Anti-RPL14 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RPL14 (ribosomal protein L14)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human