Anti-RRAS Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3727AEA 350 EUR
ABGEAT3727A
Anti-RRAS Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RRAS (related RAS viral (r-ras) oncogene homolog)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR