Anti-SKAP1 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT3782AEA 350 EUR
ABGEAT3782A
Anti-SKAP1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SKAP1 (src kinase associated phosphoprotein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human