Anti-SCGB1A1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3785AEA 350 EUR
ABGEAT3785A
Anti-SCGB1A1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SCGB1A1 (secretoglobin, family 1A, member 1 (uteroglobin))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR