Anti-SEMA7A Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3809AEA 350 EUR
ABGEAT3809A
Anti-SEMA7A Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SEMA7A (semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR