Anti-SERPINA6 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT3819AEA 350 EUR
ABGEAT3819A
Anti-SERPINA6 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SERPINA6 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 6)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human