Anti-SERPINE1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3824AEA 350 EUR
ABGEAT3824A
Anti-SERPINE1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SERPINE1 (serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR