Anti-SF3A2 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3836AEA 350 EUR
ABGEAT3836A
Anti-SF3A2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SF3A2 (splicing factor 3a, subunit 2, 66kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR