Anti-SGK2 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT3856AEA 350 EUR
ABGEAT3856A
Anti-SGK2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SGK2 (serum/glucocorticoid regulated kinase 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human