Anti-SH3D19 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3872AEA 350 EUR
ABGEAT3872A
Anti-SH3D19 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SH3D19 (SH3 domain containing 19)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR