Anti-SHC1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3880AEA 350 EUR
ABGEAT3880A
Anti-SHC1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SHC1 (SHC (Src homology 2 domain containing) transforming protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR