Anti-SLC25A6 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3913AEA 350 EUR
ABGEAT3913A
Anti-SLC25A6 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SLC25A6 (solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 6)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR