Anti-SLC2A4RG Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3920AEA 350 EUR
ABGEAT3920A
Anti-SLC2A4RG Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SLC2A4RG (SLC2A4 regulator)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR