Anti-SLC35A3 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3922AEA 350 EUR
ABGEAT3922A
Anti-SLC35A3 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SLC35A3 (solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member A3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR