Anti-SLC9A1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3932AEA 350 EUR
ABGEAT3932A
Anti-SLC9A1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SLC9A1 (solute carrier family 9, subfamily A (NHE1, cation proton antiporter 1), member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR