Anti-SMARCD3 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3955AEA 350 EUR
ABGEAT3955A
Anti-SMARCD3 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SMARCD3 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR