Anti-SMPDL3B Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT3963AEA 350 EUR
ABGEAT3963A
Anti-SMPDL3B Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SMPDL3B (sphingomyelin phosphodiesterase, acid-like 3B)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR