Anti-TNFSF12 Mouse Monoclonal Antibody

Lieferant: Abgent

ABGEAT4285AEA 350 EUR
ABGEAT4285A
Anti-TNFSF12 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TNFSF12 (tumor necrosis factor (ligand) superfamily, member 12)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR