Anti-TOMM22 Mouse Monoclonal Antibody

Lieferant: Abgent

ABGEAT4302AEA 350 EUR
ABGEAT4302A
Anti-TOMM22 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TOMM22 (translocase of outer mitochondrial membrane 22 homolog (yeast))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR