Anti-TOP3B Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT4307AEA 350 EUR
ABGEAT4307A
Anti-TOP3B Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TOP3B (topoisomerase (DNA) III beta)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human