Anti-TREM1 Mouse Monoclonal Antibody

Lieferant: Abgent

ABGEAT4336AEA 350 EUR
ABGEAT4336A
Anti-TREM1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TREM1 (triggering receptor expressed on myeloid cells 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR