Anti-TSC22D4 Mouse Monoclonal Antibody

Lieferant: Abgent

ABGEAT4375AEA 350 EUR
ABGEAT4375A
Anti-TSC22D4 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TSC22D4 (TSC22 domain family, member 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR