Anti-UBE2N Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT4441AEA 350 EUR
ABGEAT4441A
Anti-UBE2N Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: UBE2N (ubiquitin-conjugating enzyme E2N)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR