Anti-UCHL3 Mouse Monoclonal Antibody

Lieferant: Abgent

ABGEAT4456AEA 350 EUR
ABGEAT4456A
Anti-UCHL3 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: UCHL3 (ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR