Anti-UMPS Mouse Monoclonal Antibody

Lieferant: Abgent

ABGEAT4467AEA 350 EUR
ABGEAT4467A
Anti-UMPS Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: UMPS (uridine monophosphate synthetase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR