Anti-VAMP1 Mouse Monoclonal Antibody

Lieferant: Abgent

ABGEAT4500AEA 350 EUR
ABGEAT4500A
Anti-VAMP1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: VAMP1 (vesicle-associated membrane protein 1 (synaptobrevin 1))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR