Anti-VPS18 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT4520AEA 350 EUR
ABGEAT4520A
Anti-VPS18 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: VPS18 (vacuolar protein sorting 18 homolog (S. cerevisiae))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR